Learn More
Description
- Mouse keratinocyte growth factor (KGF) consists of amino acids 32 to 194
- Expressed in E. coli
- Mature mouse KGF amino acid sequence is 96% identical with rat, bovine, ovine, equine, and human forms
- Active across species
Specifications
Specifications
| Formulation | Lyophilized from 0.2μm sterile-filtered MOPS, Na2SO4, EDTA, DTT BSA. |
| Quantity | 25 μg |
| Storage Requirements | -20°C |
| Endotoxin Concentration | >1.0 EU per/1μg protein |
| Product Type | Recombinant Mouse KGF/FGF-7 Protein |
| Form | Solid |
| Purity or Quality Grade | ≥90% by SDS-PAGE |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.