Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Serine/Threonine Kinase Inhibitor Set

Catalog No. 5395721SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-957-21SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-957-21SET Supplier MilliporeSigma™ Supplier No. 5395721SET

The Serine/Threonine Kinase Inhibitor Set controls the biological activity of Serine/Threonine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Hygroscopic
Form Solid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.