Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT5 Inhibitor

Catalog No. 57310810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-310-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-310-810MG Supplier MilliporeSigma™ Supplier No. 57310810MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable compound that selectively targets SH2 domain of STAT5

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Purity 97% by HPLC
Content And Storage Protect from Light
Quantity 10 mg
Product Type STAT5 Inhibitor
Molecular Weight (g/mol) 293.3g/mol
Form Solid

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.