Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TGF-β1, Human Platelets

Catalog No. 6164501UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-645-01UG 1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-645-01UG Supplier MilliporeSigma™ Supplier No. 6164501UG

Promotes apoptosis in resting human B lymphocytes, glioma cells trigeminal neurinomal cells.

EMD Millipore Calbiochem™ TGF-β1, Human Platelets are native transforming growth factor-β1 from human platelets.

  • Contaminants
  • Endotoxin: ≤1 EU/μg of TGF-β1
  • ED₅₀ = 40 to 10pg/mL, measured by inhibition of IL-4-dependant proliferation of mouse HT-2 cells using resazurin dye as an indicator of cell proliferation
  • Lyophilized from sterile-filtered solution of 50μg BSA/μg TGF-β1, 35% Acetonitrile, 0.1% TFA
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 μg
Assay ≥97% by SDS-PAGE
Synonym hTGF-β1, Transforming Growth Factor-β1
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.