Learn More
MilliporeSigma™ Calbiochem™ Thyroid Stimulating Hormone, Human Pituitary, Iodination Grade
Shop All MilliporeSigma ProductsDescription
- Secretion is inhibited by Somatostatin
Specifications
Specifications
| Quantity | 100 μg |
| Source | Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. |
| Storage Requirements | -20°C |
| Common Name | Thyrotropin, hTSH |
| Product Type | Thyroid Stimulating Hormone, Human Pituitary |
| Form | Lyophilized |
| Purity or Quality Grade | ≥95% by weight |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.