Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Thyroid Stimulating Hormone, Human Pituitary, Iodination Grade

Catalog No. 86900650UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-900-650UG 50 μg
86-900-610UG 10 μg
86-900-6100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 86-900-650UG Supplier MilliporeSigma™ Supplier No. 86900650UG
Add to Cart
Add to Cart

Glycoprotein hormone that exerts mild, continuous stimulation on thyroid, resulting in maintenance of activity

  • Secretion is inhibited by Somatostatin
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg
Source Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Storage Requirements -20°C
Common Name Thyrotropin, hTSH
Product Type Thyroid Stimulating Hormone, Human Pituitary
Form Lyophilized
Purity or Quality Grade ≥95% by weight

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.