Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TLR1/TLR2 Antagonist, CU-CPT22

Catalog No. 61430510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-430-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-430-510MG Supplier MilliporeSigma™ Supplier No. 61430510MG
Add to Cart
Add to Cart

A cell-permeable benzotropolone compound that competes against Pam3 CSK4

A cell-permeable benzotropolone compound that competes against Pam3CSK4- (Cat. No. 506350) for TLR1/TLR2 heterodimer binding ( K i = 410nM) and selectively antagonizes against Pam3CSK4-induced RAW264.7 cellular NO production (IC50 = 580nM) without apparent cytotoxicity ( up to 100μM and 24 h). Exhibits no antagonist activity toward TLR2/TLR6, TLR3, TLR4, TLR7 at 0.5μM and little inhibitory potency against a panel of 10 kinases at 5μM (by <11% inhibition).
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 98% by HPLC
Target Proteins TLR1
Molecular Formula C19H22O7
Formula Weight 362.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.