Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TMB, Insoluble

Catalog No. 613548100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-354-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-354-8100ML Supplier MilliporeSigma™ Supplier No. 613548100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peroxidase substrate which produces stable, insoluble, aquamarine blue precipitate at reaction site.

  • Peroxidase substrate
  • Produces precipitate at reaction site with little or no background
  • CAS: 54827-17-7
  • Color: Clear to Light Brown
TRUSTED_SUSTAINABILITY

Specifications

Product Type TMB Substrate
Quantity 100 mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.