Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TNF-α Antagonist

Catalog No. 6542551MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-425-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-425-51MG Supplier MilliporeSigma™ Supplier No. 6542551MG
Add to Cart
Add to Cart

The TNF-α Antagonist controls the biological activity of TNF-α.

Peptide Sequence: H-Tyr-Cys-Trp-Ser-Gln-Tyr-Leu-Cys-Tyr-OH (disulfide bond: 2 → 8)
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins TNF-α
Molecular Formula C58H71N11O15S2
Formula Weight 1226.4g/mol
Solubility H2O or 5% Acetic Acid
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.