Learn More
Lascar Electronics Inc NIST Calibration Certificate for EasyLog Data Loggers
SDP
Supplier:  Lascar Electronics Inc CALCERTELUSB1/0DEG
Encompass
This is a single-point, NIST traceable temperature calibration test certificate available for any of Lascar's temperature data loggers (except EL-USB-RT). Generated in Lascar’s ISO 17025 calibration facility, each test certificate is valid for 12 months and indicates that the purchased data logger has been tested at 0°C /+32°F and conforms to the stated specification.
Catalog No. NC1751285