Learn More
Novus Biologicals™ Calcein, AM Solution

Description
Biological Activity: Cell permeable, non fluorescent compound; hydrolyzed by intracellular esterases to become fluorescent calcein in living cells. Used for monitoring cell viability, chemotaxis, cell adhesion and multidrug resistance.
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Formulation | C46H46N2O23 |
| For Use With (Application) | Fluorescence monitoring |
| Quantity | 1 mg |
| Product Type | Fluorescein complex |
| Molecular Weight (g/mol) | 994.86g/mol |
| Solubility | Soluble to 100mM in DMSO |
| Content And Storage | -20°C |
| Target | Calcein |
| Color | White |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.