Learn More
Biotium Calcein (High Purity)
Supplier: Biotium 80013
Calcein is a highly negatively charged, water-soluble fluorescein derivative. Its fluorescence is nearly independent of pH in the range between 6.5 and 12. The dye can be introduced into cells by microinjection and is useful for cell tracing and for the study of cell membrane integrity and fusion. Biotium also offers calcein AM, the membrane-permeant version of calcein that hydrolyzes into calcein once inside cells. Properties: Ex/Em = 494/517 nm (pH 8). e = 75,000 Yellow-orange solid soluble in DMSO, DMF or pH >6 water; Store at 4°C and protect from light; C30H26N2O13; MW: 623; [154071-48-4].
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.