Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcein, MP Biomedicals™

Catalog No. ICN19016701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19016701 1 g
ICN19016705 5 g
ICN19016710 10 g
ICN19016725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19016701 Supplier MP Biomedicals, Inc Supplier No. 0219016701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1461-15-0
Molecular Formula C30H26N2O13
Molecular Weight (g/mol) 622.539
InChI Key DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Fluorexon, Calcein W
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Formula Weight 622.5
Color Orange
Quantity 1 g
Physical Form Powder
Chemical Name or Material Calcein, Suitable for fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.