Learn More
Apexbio Technology LLC BS-181 1092443-52-1 25mg
SDP
Supplier:  Apexbio Technology LLC A326625
Encompass_Preferred
BS-181 (CAS 1092443-52-1) is a small molecule inhibitor developed to selectively target cyclin-dependent kinase 7 (CDK7) a regulator of cell cycle progression In in vitro kinase assays BS-181 demonstrates a 35-fold selectivity for CDK7 over CDK2 with an IC50 of 880 nM for CDK7 BS-181 inhibits phosphorylation of CDK7 substrates in MCF-7 cancer cells resulting in cell cycle arrest and induction of apoptosis thereby suppressing cancer cell proliferation In vivo studies show that BS-181 is stable following intraperitoneal administration and reduces tumor growth in MCF-7 xenograft models BS-181 serves as a useful tool for probing CDK7 function and potential anti-tumor effects in preclinical research
Catalog No. 50-000-92880
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.