Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calciferol 98.0+%, TCI America™
SDP

Catalog No. C00055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00055G 5 g
C00051G 1 g
C000525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C00055G Supplier TCI America Supplier No. C00055G

Chemical Identifiers

CAS 50-14-6
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.66
MDL Number MFCD00166988,MFCD11656674
InChI Key MECHNRXZTMCUDQ-VLOQVYPSSA-N
Synonym ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
PubChem CID 134129658
IUPAC Name (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

Melting Point 114°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 396.66
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Calciferol
RTECSNumber KE1050000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.