Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Calcined petroleum coke powder, crystalline, -325 mesh, 99%

Catalog No. AA4727530
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4727530 250 g
AA4727514 25 g
AA47275A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4727530 Supplier Thermo Scientific™ Supplier No. 4727530
Add to Cart

CAS: 64743-05-1

TRUSTED_SUSTAINABILITY

Specifications

CAS 64743-05-1
Density 2.1
Quantity 250 g
Physical Form Solid
Synonym Coke (petroleum), calcined;
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.