Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcineurin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674127
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-127 Supplier Invitrogen™ Supplier No. PA112394
  • $659.00
    $547.20 / Each of 1

    Save $111.80

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcineurin A Polyclonal Antibody for Western Blot, IHC

Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. Alternative splicing results in multiple transcript variants encoding two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PPP3CA
Gene Accession No. P48452, P63328, P63329, Q08209
Gene Alias 2900074D19Rik; Ca2+/calmodulin-regulated protein phosphatase; calcineurin; calcineurin A alpha; Calcineurin A1; calcineurin catalytic subunit; calcineurin catalytic subunit delta isoform; Calcineurin subunit A alpha; CalcineurinA; calmodulin-dependent calcineurin A subunit alpha isoform; caln; CALNA; CALNA 1; CALNA1; CAM-PRP catalytic subunit; CCN 1; ccn1; CN; CnA; CNA alpha; CNA1; PP2B alpha 1; PP2BA alpha; PP2CB; ppp2b; ppp3ca; ppp3ca.S; protein phosphatase; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform (calcineurin A alpha); protein phosphatase 3 catalytic subunit alpha; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isoform; protein phosphatase 3, catalytic subunit, alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme S homeolog; protein phosphatase 3, catalytic subunit, gamma isozyme; protein phosphatase-2Ba; serine/threonine protein kinase; serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform; XELAEV_18009418mg
Gene Symbols PPP3CA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues R(482) M P P R R D A M P S D A(494) of protein phosphatase 2B alpha.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19055, 24674, 286852, 5530
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.