Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Calcineurin B Protein
SDP

Catalog No. NBP17879710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17879710 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17879710 Supplier Novus Biologicals™ Supplier No. NBP178797100UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-170 of Human Calcineurin B The Recombinant Human Calcineurin B Protein is derived from E. coli. The Recombinant Human Calcineurin B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000936
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl.
Gene ID (Entrez) 5534
Molecular Weight (g/mol) 21.5kDa
Purification Method SDS-PAGE
Quantity 100μg
Source Recombinant human PPP3R1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen PPP3R1, 1-170aa, Human, His tag, E. coli (NP_000936).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Calcineurin B
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.