Learn More
Genscript Corporation Calcitonin 12C5 mAb Mouse
Supplier:  Genscript Corporation V0190210
Encompass_Preferred
PCT a 116 amino acid (aa) protein is comprised of three sections including a 57 aa N-terminal PCT a 32 aa calcitonin and a 21 aa katacalcin Calcitonin is a hormone derived from PCT cleavage PCT is a good diagnosis marker for bacterial infection Other diseases such as sepsis inflammation surgery heat shock burn injuries and cardiogenic shock can also cause an increase of PCT level in blood
Catalog No. 50-000-59036
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.