Learn More
Biocare Medical, LLC Calcitonin Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PP072AA
Encompass_Preferred
Studies have shown that calcitonin antibody reacts with the human protein calcitonin and labels C-cells in normal thyroid. Calcitonin antibody has been reported to be particularly useful in differentiating medullary carcinoma from papillary and follicular thyroid cancer. Most medullary carcinomas are positive for calcitonin; conversely, most papillary and follicular types of thyroid cancer are usually negative for calcitonin. When used in conjunction with TTF-1 thyroid medullary carcinoma may be distinguishable from laryngeal moderately differentiated carcinoma.
Catalog No. 50-831-95
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.