Learn More
CPC Scientific H-Cys-Gly-Asn-Leu-Ser-Thr-Cys-Met-Leu-Gly-Thr-Tyr-Thr-Gln-Asp-Phe-Asn-Lys-Phe-His-Thr-Phe-Pro-Gln-Thr-Ala-Ile-Gly-Val-Gly-Ala-Pro-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific OSTP014A
Encompass
Inhibitor of gastrin and gastric acid secretion that leads to a lowering of plasma calcium and phosphate levels by inhibiting bone reabsorption and excretion through the kidneys.SEQUENCE: H-Cys-Gly-Asn-Leu-Ser-Thr-Cys-Met-Leu-Gly-Thr-Tyr-Thr-Gln-Asp-Phe-Asn-Lys-Phe-His-Thr-Phe-Pro-Gln-Thr-Ala-Ile-Gly-Val-Gly-Ala-Pro-NH2 (trifluoroacetate salt)(Cys1 and 7 bridge)ONE-LETTER SEQUENCE: CGNLSTCMLGTYTQDFNKFHTFPQTAIGVGAP-NH2 (C1&C7 bridge)MOLECULAR FORMULA: C151H226N40O45S3MOLECULAR WEIGHT: 3417.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [21215-62-3]SYNONYMS: Calcitonin M, human C carcinoma, Calcitonin, human reduced cyclic (1-7)-disulfide, Thyrocalcitonin, hCTRESEARCH AREA: OsteoporosisREFERENCES:Y. Nakagawa, et al., Peptide Chemistry, 1976, 189 (1977)SKU(s): OSTP-014A, OSTP-014B
Catalog No. 50-211-0023
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.