Learn More
Invitrogen™ Human Procalcitonin Recombinant Protein
Description
RP-75697 contains a purified 114 amino acid fragment (aa3-116) corresponding to the unprocessed human procalcitonin protein with an N-terminal hexahistidine tag. RP-75697 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P01258, P06881 |
| Concentration | 0.1 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8.0 |
| Gene ID (Entrez) | 796 |
| Name | Human Procalcitonin |
| pH Range | 8.0 |
| Quantity | 10 μg |
| Source | E. Coli |
| Immunogen | N/A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.