Learn More
CPC Scientific H-Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 5MG
SDP
Supplier:  CPC Scientific OSTP012C
Encompass
This peptide has the ability to cross mucous membranes with no significant effect on inositol phosphate accumulation.SEQUENCE: H-Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2(Cys1 and 7 bridge)ONE-LETTER SEQUENCE: CSNLSTCVLGKLSQELHKLQTYPRTNTGSGTP-NH2 (C1&C7 bridge)MOLECULAR FORMULA: C145H240N44O48S2MOLECULAR WEIGHT: 3431.90STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [47931-85-1]SYNONYMS: Salcatonin, Salmon Calcitonin, Thyrocalcitonin (salmon)RESEARCH AREA: OsteoporosisREFERENCES:C.R. Hamilton, Am. J. Med., 56, 858 (1974)
SKU(s): OSTP-012A, OSTP-012B, OSTP-012C, OSTP-012D
Catalog No. 50-211-0019
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.