Learn More
Abcam Calcitriol, Vitamin D Receptor Agonist, Solid, 250 μg
Description
- Activity: Vitamin D receptor (VDR) agonist
- Purity: >99%
- Molecular Weight: 416.6 Da
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Vitamin D Receptor (VDR) |
| Form | Solid |
| Formulation | 100% Calcitriol |
| Quantity | 250 μg |
| For Use With (Application) | Vitamin D receptor (VDR) agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.