Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium
SCI_ED

Catalog No. S25217B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Reagent Grade - Granular
Reagent Grade - Turnings
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25217B 500 g Reagent Grade - Turnings
S25216 25 g Reagent Grade - Granular
S25217 25 g Reagent Grade - Turnings
S25217A 100 g Reagent Grade - Turnings
S25216A 500 g Reagent Grade - Granular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S25217B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reacts with water or moisture

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Silver-White
Density 1.54g/mL at 20°C/4°C
Quantity 500 g
Packaging Reagent Grade - Turnings
Linear Formula Ca
Formula Weight 40.08
Chemical Name or Material Calcium
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 4.3, II, : Dangerous When Wet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.