Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium acetate monohydrate, ACS, 99.0% min

Catalog No. p-4407308
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1344630 250 g
AA13446A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1344630 Supplier Thermo Scientific Chemicals Supplier No. 01344630
Only null left
Add to Cart

CAS: 5743-26-0 | C4H8CaO5 | 176.181 g/mol

Calcium acetate monohydrate is used to precipitate oxalates in solution. It is used to control the phosphate levels in blood, thereby controlling the bone problems. It is also involved in the neutralization of fluoride in water. Further, it is used as a food additive mainly in candy products. In addition to this, it serves as a stabilizer, buffer and sequestrant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.181
MDL Number MFCD00150019
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99%
Beilstein 3692527
Solubility Information Soluble in water. Slightly soluble in methanol and hydrazine. Insoluble in ethanol,acetone and benzene.
Formula Weight 176.18 (158.17 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 1.5 g/cm3
Chemical Name or Material Calcium acetate monohydrate
Show More Show Less
Hazard Category H303: May be harmful if swallowed.
EINECSNumber 200-540-9
RTECSNumber AF7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.