Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Acetate Monohydrate (Powder/Certified), Fisher Chemical

Catalog No. c46500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C46-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. C46-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.181
MDL Number MFCD00150019
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Melting Point 160°C
Color White
pH 7 to 8
Physical Form Solid
Quantity 500 g
Linear Formula Ca(CH3COO)2·H2O
Formula Weight 176.18
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Decomposition Information 160°C
Chemical Name or Material Calcium Acetate Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability:
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.