Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Calcium Assay Buffer, 15 mL

Catalog No. 502704421
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4421 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4421 Supplier Abcam Supplier No. AB18245815ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buffer optimized for calcium measurement in assay kits.

Calcium Assay Buffer designed for the measurement of calcium. This product is cited in 1 publication.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Wash Solution
Format Bottle
Form Liquid
Target Molecule Calcium
Quantity 15 mL
For Use With (Application) Calcium measurement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.