Learn More
Abcam Calcium Assay Kit, Luminometric, Cell-based, 100x96 Tests
Description
- Assay type: Cell-based
- Detection method: Luminescent
- Sample types: Suspension cells, Adherent cells
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| For Use With (Equipment) | Microplate Reader |
| No. of Reactions | 9600 Tests |
| Detection Location | In-Solution Detection |
| Substrate Properties | Chemical Substrate |
| Detection Method | Chemiluminescence |
| Format | Kit |
| Target Molecule | Calcium |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 9,600 Tests |
| For Use With (Application) | Calcium measurement in cell samples |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.