Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Carbonate
SCI_ED

Catalog No. S25220A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Natural Chalk - Solid
Reagent Grade - Solid
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25220A 500 g Reagent Grade - Solid
S25220 100 g Reagent Grade - Solid
S25220B 500 g Natural Chalk - Solid
S25220C 2.5 kg Reagent Grade - Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S25220A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CO2 generation

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Color White
Quantity 500 g
Assay Percent Range 100% w/w
Linear Formula CaCO3
Formula Weight 100.09
Packaging Reagent Grade - Solid
Density ∽2.7 to 2.9g/mL
Chemical Name or Material Calcium Carbonate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.