Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, ACS Chelometric Standard, Reagents

Catalog No. 01338067
Change view
Click to view available options
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-067 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-067 Supplier Reagents Holdings Llc Supplier No. C1161550100C2
Add to Cart
Add to Cart

Calcium Carbonate, ACS Chelometric Standard, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym Limestone, Marble, Chalk, Aragonite, Calcite
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825°C
Color White
pH 8 to 9
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.83
Chemical Name or Material Calcium Carbonate, ACS Chelometric Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.