Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCC0050500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Natural Poly Bottle
Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCC0050500 500 g Poly Wide Mouth Bottle
RDCC0050125 125 g Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCC0050500 Supplier Ricca Chemical Company Supplier No. RDCC0050500B1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Powder
Quantity 500 g
Percent Purity ≥ 99%
Grade ACS
Packaging Poly Wide Mouth Bottle
Chemical Name or Material Calcium Carbonate
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.