Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Heavy Powder, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601222 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
45 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-222 25 kg Fiber Drum
18-601-217 500 g Poly Bottle
18-601-218 2.5 kg Poly Pail
18-601-219 12 kg Poly Pail
18-601-221 45 kg Fiber Drum
18-601-220 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-222 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA13125KGBL

missing translation for 'validMainframeMsg'

CA131, 471-34-1, CaCO3

Spectrum™ Chemical Calcium Carbonate, Heavy Powder, USP is used as a pharmaceutical excipient solid dosage diluent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 25 kg
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 2%
Packaging Fiber Drum
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.