Learn More
Calcium carbonate, ACS, low in alkalies, 99.0% min, Thermo Scientific Chemicals
Description
It is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles shows an useful application as anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 800°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 99.0% min. |
Solubility Information | Soluble in acids,insoluble in water. |
Formula Weight | 100.09 |
Grade | ACS |
Refractive Index | 1.6583 |
Density | 2.93 |
Show More |
Safety and Handling
EINECSNumber | 207-439-9 |
RTECSNumber | FF9335000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.