Learn More
Description
A suggested positive control for this product is rat brain lysate.
Specifications
Specifications
| Antigen | Calcium Channel beta-4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Cacnb4 |
| Gene Accession No. | O00305 |
| Gene Alias | 3110038O15Rik; CAB4; Cacnb4; CACNLB4; calcium channel beta 4 subunit; calcium channel voltage-dependent subunit beta 4; calcium channel, voltage-dependent, beta 4 subunit; calcium voltage-gated channel auxiliary subunit beta 4; Cchb4; dihydropyridine-sensitive L-type, calcium channel beta-4 subunit; EA5; EIG9; EJM; EJM4; EJM6; lh; OTTHUMP00000207247; OTTHUMP00000207249; Voltage-dependent L-type calcium channel subunit beta-4 |
| Gene Symbols | Cacnb4 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
