Learn More
Calcium Chloride, 0.100 M (M/10), 0.200 N (N/5), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| CAS Min % | 97.0, 1.42 |
| CAS Max % | 100.0, 1.48 |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 0.100 Molar (M/10), 0.200 Normal (N/5) |
| Packaging | Natural Poly Bottle |
| pH | 8 to 9 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Chemical Name or Material | Calcium Chloride |
Safety and Handling
| ShelfLife | 18 Months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.