Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, 0.500 M (M/2), Ricca Chemical
SDP

Catalog No. R176400020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R176400020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R176400020F Supplier Ricca Chemical Company Supplier No. R176400020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10035-04-8 , 7732-18-5

Specifications

Color Colorless
Grade Laboratory
Odor Odorless
Packaging Cubitainer
pH 8 to 9
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Calcium Chloride, 0.500 Molar (M/2)
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.