Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, 10% (w/v), Ricca Chemical
SDP

Catalog No. 172232 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
172232 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 172232 Supplier Ricca Chemical Company Supplier No. 172232
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 10035-04-8
Molecular Weight (g/mol) 110.98 g/mol

Specifications

Physical Form Liquid
Quantity 1 L
Percent Purity 10% W/V
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Calcium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.