Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride 2-Hydrate, DPL

Catalog No. 502985583
Encompass
Catalog No. 50-298-5583 Supplier Thermo Scientific Production Chemicals and Services Supplier No. D51103001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10035-04-8
Molecular Weight (g/mol) 147 g/mol

Specifications

Physical Form Crystalline
Quantity 10 kg
Percent Purity 97.0 to 103.0% (Ph.Eur.) 99.0 to 107.0% (USP)
Grade USP
Packaging Plastic container
Chemical Name or Material Calcium chloride dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.