Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride, Anhydrous, powder, ≥97%, Solstice

Catalog No. 60047905 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-905 2.5 kg Poly Bottle
60-047-904 100 g Poly Bottle
60-047-906 500 g Poly Bottle
600-30-26 1 kg Poly Bottle
60-047-907 5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-047-905 Supplier Solstice Supplier No. C49012.5KG
Add to Cart
Add to Cart

Anhydrous, powder,≥97%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point 1,900°C
Melting Point 772°C (literature)
CAS Max % 100.0000%
Physical Form Powder
Quantity 2.5 kg
Linear Formula CaCl2
UN Number NONH for all modes of transport
IUPAC Name calcium;dichloride
Formula Weight 110.98
Percent Purity >97%
Packaging Poly Bottle
Vapor Pressure 0.01 mmHg (20°C)
Density 2.150 g/cm3
Chemical Name or Material Calcium chloride, Anhydrous, powder, ≥97%
Show More Show Less
Hazard Statement P305 + P351 + P338
ShelfLife 1620 days from date of manufacture
EINECSNumber 233-140-8
RTECSNumber EV9800000
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.