Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Certified, 0.0200M ±0.0002M (0.04N), LabChem™

Catalog No. LC128001 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC128001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC128001 Supplier LabChem, Inc. Supplier No. LC128001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10035-04-8 , 7732-18-5

Specifications

Color Colorless
CAS Max % 99.71, 0.29
Physical Form Liquid
Quantity 500 mL
Linear Formula CaCl2·2H2O
Solubility Information Soluble in water
Formula Weight 147.02
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0200M ±0.0002M (0.04N)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride
Chemical Name or Material Calcium Chloride
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.