Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Chloride, Dihydrate
SCI_ED

Catalog No. S25221A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Flakes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25221A 500 g Lab Grade - Flakes
S25221 100 g Lab Grade - Flakes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25221A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O

Specifications

Color White
Quantity 500 g
Assay Percent Range >95% w/w
Linear Formula CaCl2·2H2O
Formula Weight 147.02
Packaging Lab Grade - Flakes
Density 0.83g/mL
Chemical Name or Material Calcium Chloride, Dihydrate, Dihydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.