Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, ACS Grade, 99.0 to 105.0%, LabChem™
GSA_VA

Catalog No. LC127251 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC127251 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC127251 Supplier LabChem, Inc. Supplier No. LC127251
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 10035-04-8

Specifications

Color White
CAS Max % 100
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula CaCl2·2H2O
Solubility Information Soluble in water
Formula Weight 147.02
Percent Purity 99.0 to 105.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 0.835g/cm3
Decomposition Information Hydrogen chloride
Density 0.835g/cm3
Chemical Name or Material Calcium Chloride, Dihydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 233-140-8
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.