Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCC0075500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
25 kg
Packaging:
Fiber Drum
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCC0075500 500 g White Poly Wide Mouth Bottle
RDCC007525F 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCC0075500 Supplier Ricca Chemical Company Supplier No. RDCC0075500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10035-04-8
Molecular Weight (g/mol) 147.01 g/mol

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Granular/Lump
Quantity 500 g
Percent Purity 99 to 105%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Calcium Chloride Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.