Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, ACS Reagent, Solstice

Catalog No. 6002153 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-021-53 6 x 500 g
60-021-52 500 g
60-021-49 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-021-53 Supplier Solstice Supplier No. 2235066X500G/DEL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
MDL Number MFCD00149613

Specifications

Quantity 6 x 500 g
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99%
Grade ACS Reagent
Vapor Pressure 0.01mmHg (20°C)
Chemical Name or Material Calcium Chloride Dihydrate
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 233-140-8
RTECSNumber EV9810000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.