Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, EP, BP, JP, USP, 99-103%, Spectrum™ Chemical
SDP

Catalog No. 18601275 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-275 50 kg Fiber Drum
18-601-272 500 g Poly Bottle
18-601-273 2.5 kg Poly Bottle
18-601-274 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-275 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA14050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CA140, 10035-04-8, CaCl2·2H2O

Spectrum™ Chemical Calcium Chloride, Dihydrate, USP, EP, BP, JP is used in treatment of hypocalcemia, electrolyte depletion, and hyperkalemia. All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2H4O2
Molecular Weight (g/mol) 147.01
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
IUPAC Name calcium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ca++]

Specifications

pH 4.5-9.2
CAS Max % 100%
Quantity 50 kg
Percent Purity 99 to 103%
Grade Multi-Compendial
Packaging Fiber Drum
Appearance Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.