Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calcium Chloride, Dihydrate OmniPur™ Calbiochem™,

Catalog No. 3000500GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-005-00GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-005-00GM Supplier MilliporeSigma™ Supplier No. 3000500GM

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10035-04-8
Synonym Calcium Chloride

Specifications

Quantity 500 g
Assay Percent Range 99.0 to 105.0%
Formula Weight 147
Chemical Name or Material Calcium Chloride, Dihydrate
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.