Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, Reagent, ACS, 99-105%, Spectrum™ Chemical
SDP

Catalog No. 18601265 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-265 125 g Poly Bottle
18-601-266 500 g Poly Bottle
18-601-268 2.5 kg Poly Bottle
18-601-267 6 x 500 g Poly Bottle
18-601-269 4 x 2.5 kg Poly Bottle
18-601-270 12 kg Poly Pail
18-601-271 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-601-265 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1096125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1096, 10035-04-8, CaCl2·2H2O

Spectrum™ Chemical Calcium Chloride, Dihydrate, Reagent, ACS is a analytical grade chemical that is packaged in laboratory size bottles to production drum quantities. As an ACS grade Reagent, Spectrum Chemical manufactured Calcium Chloride is used as the quality standard against which other Calcium Chloride substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2H4O2
Molecular Weight (g/mol) 147.01
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
IUPAC Name calcium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ca++]

Specifications

pH 4.5- 8.5
CAS Max % 100%
Quantity 125 g
Percent Purity 99 to 105%
Grade ACS
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.