Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Standard, 1000 ppm CaCO3 (400 ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+, Ricca Chemical
SDP

Catalog No. 178032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1780-32 1 L Natural Poly Bottle
17804 120 mL Natural Poly Bottle
1780-16 500 mL Natural Poly Bottle
R17800004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 1780-32 Supplier Ricca Chemical Company Supplier No. 178032
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 471-34-1 , 7647-01-0 , 7732-18-5
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.19, 0.1
CAS Max % 100.0, 0.2, 0.1
Equivalence 1mL= 1 mg CaCO3
Boiling Point 100°C
Quantity 1 L
Concentration or Composition (by Analyte or Components) 1000ppm CaCO3 (400ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+
Chemical Name or Material Calcium Chloride Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.