Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Technical, Spectrum™ Chemical
SDP

Catalog No. 18601276 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-276 500 g Poly Bottle
18-601-277 2.5 kg Poly Bottle
18-601-278 12 kg Poly Pail
18-601-279 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-276 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1098500GM
Add to Cart
Add to Cart

C1098, 10035-04-8, CaCl2·2H2O

Spectrum™ Chemical Calcium Chloride, Technical is a salt that is made using chlorine and calcium. It is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and because of this it must be kept in tightly sealed, air tight containers. Calcium chloride has many uses. Its main use is as a desiccant and deicer. However, calcium chloride also is used as a food additive to add a salty flavor without increasing sodium content, used in medicine to treat hydrofluoric burns, as a sterilization agent for male animals, and as a source for calcium ions. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2H4O2
Molecular Weight (g/mol) 147.01
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
IUPAC Name calcium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 100%
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.